Order Entry
United States
ContactUsLinkComponent
Total Bile Acid Assay Kits, Cell Biolabs
Total Bile Acid Assay Kits, Cell Biolabs
Supplier:  Cell Biolabs
Certificates
undefined
Total Bile Acid Assay Kits, Cell Biolabs
Supplier:  Cell Biolabs

Specifications

  • Storage Temperature:
    –80°C
  • Sensitivity:
    1 µM

Specifications

Related Information

Frequently Bought Together