ContactUsLinkComponent
rh FGF Basic LifeFactor, Lifeline Cell Technology
rh FGF Basic LifeFactor, Lifeline Cell Technology
Catalog # 102970-974
CAS Number:  
rh FGF Basic LifeFactor, Lifeline Cell Technology
Catalog # 102970-974
Supplier Number:  LS-1002
CAS Number:  
Specifications
  • Description:
    rh FGF Basic LifeFactor
  • Size:
    0.5 mL
  • Cat. No.:
    102970-974
  • Supplier no.:
    LS-1002
Specifications
About this item
FGF Basic LifeFactor is used with fibroblasts, endothelial cells and smooth muscle cells.
  • Used with human fibroblasts
  • Used with human endothelial cells
  • Used with human smooth muscle cells
  • Used with basal medium
FGF Basic LifeFactor may be used with these Lifeline® products: a) FibroLife® basal medium and human fibroblasts, b) VascuLife® basal medium and human endothelial cells, c) VascuLife® basal medium and human smooth muscle cells.