Specifications
- Description:HBTEC Air-Liquid Interface Differentiation Complete Frozen Medium
- Size:500 mL
- Cat. No.:102970-798
- Supplier no.:LM-0050
Specifications
About this item
HBTEC Air-Liquid Interface Differentiation Complete Frozen Medium is optimized for differentiation of human bronchial/tracheal epithelial cells.
- Air-liquid interface mucociliary differentiation
- Convenient
- Easy to use
- Fully supplemented
HBTEC Air-Liquid Interface Differentiation Complete Frozen Medium is optimized for air-liquid interface mucociliary differentiation of human bronchial/tracheal epithelial cells.
Offered as a convenient and easy to use, fully-supplemented complete medium, stored frozen to extend shelf life.