- Cell Format:Cryopreserved
- Description:Human Coronary Artery Endothelial Cells
- Size:500,000 cells per vial
- Cat. No.:102970-676
- Supplier no.:FC-0032
Lifeline® Normal Human Coronary Artery Endothelial Cells are isolated to ensure that they remain unexposed to recombinant human VEGF.
- Can be grown in low serum medium
- No exposure to recombinant human VEGF
- May be cultured with or without VEGF
Lifeline® Normal Human Coronary Artery Endothelial Cells are isolated in VascuLife® EnGS Medium to ensure that they remain unexposed to recombinant human VEGF. Coronary Artery Endothelial Cells may be cultured with or without VEGF.