ContactUsLinkComponent
OsteoLife™ Complete Medium, Lifeline Cell Technology
OsteoLife™ Complete Medium, Lifeline Cell Technology
Catalog # 102970-780
CAS Number:  
OsteoLife™ Complete Medium, Lifeline Cell Technology
Catalog # 102970-780
Supplier Number:  LM-0023
CAS Number:  
Specifications
  • Description:
    OsteoLife™ Osteogenesis Complete Frozen Medium
  • Size:
    100 ml
  • Cat. No.:
    102970-780
  • Supplier no.:
    LM-0023
Specifications
About this item
OsteoLife™ Osteogenesis Medium is optimized for the differentiation of adult human mesenchymal stem cells into osteocytes.
  • Serum-free
  • Induces differentiation of human mesenchymal stem cells into osteoblasts
  • Exhibits superior calcium deposition
  • No antimicrobials
  • No phenol red
  • Fully-supplemented frozen medium
OsteoLife™ Osteogenesis Medium is a serum-free medium optimized for the differentiation of adult human mesenchymal stem cells into osteocytes. OsteoLife™ induces the differentiation of human mesenchymal stem cells into osteoblasts that exhibit superior calcium deposition as compared with other commercially available products.
OsteoLife™ is fully supplemented and offered as a convenient and easy to use, complete frozen medium (100mL) to extend shelf-life. OsteoLife™ contains no antimicrobials and no phenol red.