ContactUsLinkComponent
Human Aortic Endothelial Cells, Lifeline Cell Technology
Human Aortic Endothelial Cells, Lifeline Cell Technology
Catalog # 102970-954
CAS Number:  
undefined
Human Aortic Endothelial Cells, Lifeline Cell Technology
Catalog # 102970-954
Supplier Number:  FC-0014
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Cell Format:
    Cryopreserved
  • Description:
    Human Aoritc Endothelial Cells
  • Size:
    500,000 cells per vial
  • Cat. No.:
    102970-954
  • Supplier no.:
    FC-0014
Specifications
About this item
Lifeline® Normal Human Aortic Endothelial Cells are quality tested and demonstrate optimal reduced-serum.
  • No exposure to antimicrobials or phenol red
  • May be cultured with or without VEGF
  • Optimal reduced-serum growth over at least 15 population doublings
Lifeline® Normal Human Aortic Endothelial Cells are quality tested in VascuLife® EnGS 2% serum medium and demonstrate optimal reduced-serum growth over a period of at least 15 population doublings at rates equal to or greater than other serum-supplemented media.
Lifeline® Aortic Endothelial Cells are not exposed to antimicrobials or phenol red when cultured in VascuLife® medium. Lifeline® Aortic Endothelial Cells may be cultured with or without VEGF.