About this item
Lifeline® Normal Human Renal Epithelial Cells are cryopreserved as Primary or Secondary cells to ensure the highest viability and plating efficiency.
- Quality tested
- Ensures optimal reduced-serum growth
- Maintenance of morphology
- No exposure to antimicrobials or phenol red
- Isolated from human kidney tissue
- Expanded in culture vessels before cryopreservation
Lifeline® normal Human Renal Epithelial Cells are quality tested in RenaLife™ Medium to ensure optimal reduced-serum growth and maintenance of morphology over a period of at least 15 population doublings at rates equal to or greater than serum-supplemented medium.
Renal Epithelial Cells can be grown in a 0.5% serum medium without phenol red or antimicrobials when cultured in RenaLife™ Medium. Lifeline® normal Human Renal Epithelial Cells are not exposed to antimicrobials or phenol red.
The cells are isolated from human kidney tissue and expanded in culture vessels before cryopreservation.