Order Entry
United States
Orders LinkContactUsLinkComponent
FrostaLife™ Cryopreservation Solution, Lifeline Cell Technology

FrostaLife™ Cryopreservation Solution, Lifeline Cell Technology

FrostaLife™ Cryopreservation Solution, 100 mL
Catalog # 102970-770
Marketsource
Supplier #:  LM-0015
FrostaLife™ Cryopreservation Solution, Lifeline Cell Technology
Catalog # 102970-770
Marketsource
CAS Number:  
FrostaLife™ Cryopreservation Solution, Lifeline Cell Technology

FrostaLife™ Cryopreservation Solution, 100 mL

CAS Number:  
Price
$246.77
Log in for availability
Each (100ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

FrostaLife™ Cryopreservation Solution is optimized for the cryopreservation of mammalian cells.

  • Xeno-Free is optimized without animal-derived components
  • Xeno-Free contains human serum
  • Xeno-Free allows freezing without inhuman component exposure
  • No antimicrobials
  • No phenol red

FrostaLife™ Cryopreservation Solution is optimized for the cryopreservation of mammalian cells. FrostaLife™ contains no antimicrobials or phenol red.

FrostaLife™ Xeno-Free is optimized for the cryopreservation of mammalian cells without the use of animal-derived components. FrostaLife™ Xeno-Free contains human serum, and allows freezing without exposure to non-human components.

Ordering information: FrostaLife™ is available as Cryopreservation Solution or as Xeno-Free (no animal-derived components) Cryopreservation Solution.

Specifications

Specifications

  • Description:
    FrostaLife™ Cryopreservation Solution
  • Size:
    100 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.