Order Entry
United States
ContactUsLinkComponent
HDL Purification Kit (Ultracentrifugation Free), Cell Biolabs
HDL Purification Kit (Ultracentrifugation Free), Cell Biolabs
Catalog # 102965-214
Supplier:  Cell Biolabs
CAS Number:  
undefined
HDL Purification Kit (Ultracentrifugation Free), Cell Biolabs
Catalog # 102965-214
Supplier:  Cell Biolabs
Supplier Number:  STA-607
CAS Number:  

Specifications

  • Description:
    HDL Purification Kit
  • Size:
    10 Preps
  • Storage Temperature:
    4°C and Room Temperature
  • Cat. No.:
    102965-214
  • Supplier no.:
    STA-607

Specifications

About this item

Traditional ultracentrifugation methods for purifying LDL, VLDL or HDL can be time consuming and inconvenient

Our HDL Purification Kit uses dextran sulfate to selectively and separately purify HDL fractions for a variety of downstream applications. No ultracentrifuge is necessary, and purification steps use lower centrifugation speeds available with most benchtop centrifuges. Each prep purifies up to 10 mL of plasma or serum, and the average yield is approximately 4.5 mg/mL of HDL.