LDL/VLDL Purification Kit (Ultracentrifugation Free), Cell Biolabs
Catalog # 102965-212
Supplier: Cell Biolabs
CAS Number:
Specifications
- Description:LDL/VLDL Purification Kit
- Size:10 Preps
- Storage Temperature:4°C and Room Temperature
- Cat. No.:102965-212
- Supplier no.:STA-606