Order Entry
United States
ContactUsLinkComponent
Gelatin Solution 0.1%, Lifeline Cell Technology
Gelatin Solution 0.1%, Lifeline Cell Technology
Catalog # 102970-804
CAS Number:  
Gelatin Solution 0.1%, Lifeline Cell Technology
Catalog # 102970-804
Supplier Number:  CM-0004
CAS Number:  

Specifications

  • Description:
    Gelatin Solution 0.1%
  • Size:
    100 mL
  • Cat. No.:
    102970-804
  • Supplier no.:
    CM-0004

Specifications

About this item

Gelatin Solution is a 0.1% solution in water of water-soluble proteins found in collagen.

  • Water-soluble proteins found in collagen

Gelatin solution is a 0.1% solution in water of water-soluble proteins found in collagen.