Accessories for Digital block reactor, DRB 200, Hach
Supplier: Hach
Certificates
About this item
This adapter converts the 20 mm diameter vial wells to 16 mm diameter.
Frequently Bought Together
This adapter converts the 20 mm diameter vial wells to 16 mm diameter.