Specifications
- Description:AdipoLife™ DfKt™-2 Adipogenesis Kit (for Wharton's Jelly, Bone Marrow MSC)
- Size:100 mL
- Cat. No.:102970-904
- Supplier no.:LL-0059
Specifications
About this item
AdipoLife™ DfKt™-2 Adipogenesis Medium is formulated to induce the adipogenic differentiation of Human Mesenchymal Stem Cells.
- Induces the adipogenic differentiation of Human Mesenchymal Stem Cells
- Derived from adipose tissue, bone marrow, and Wharton's Jelly
- No antimicrobials
- No phenol red
AdipoLife™ DfKt™-2 Adipogenesis Medium is a Lifeline® medium kit formulated to induce the adipogenic differentiation of Human Mesenchymal Stem Cells derived from adipose tissue, bone marrow, and Wharton's Jelly.
AdipoLife™ DfKt-2™ Adipogenesis Medium Complete Kit is offered in a kit format composed of AdipoLife™ Basal Medium (100mL) and a combined solution of differentiation factors called DifFactor™ 3. DifFactor™ 3 contains: rh Insulin, Indomethacin, 3-Isobutyl-1-Methylxanthine, Dexamethasone, Horse Serum (5% final conc.), L-Glutamine, and Ascorbic Acid.
AdipoLife™ DfKt™-2 contains no antimicrobials and no phenol red.