OcuLife™ Corneal Epithelial Medium Complete Kit is a Lifeline® medium optimized for the culture of human corneal epithelial cells.
- Optimized for the culture of human corneal epithelial cells
- Supports growth over at least three passages
- No antimicrobials
- No phenol red
OcuLife™ Corneal Epithelial Medium Complete Kit is a Lifeline® medium optimized for the culture of human corneal epithelial cells. OcuLife™ supports the growth of these cells over a period of at least three passages. OcuLife™ contains no antimicrobials and no phenol red.
The OcuLife™ Medium Complete Kit includes OcuLife™ Basal Medium and a Kit of the associated supplements and growth factors. The OcuLife™ LifeFactors® Kit includes: rh Insulin 0.5 mL, L-Glutamine 15 mL, Epinephrine 0.5 mL, Apo-Transferrin 0,5 mL, Extract P™ 2 mL, Hydrocortisone Hemisuccinate 0.5 mL, OcuFactor™ 1.0 mL, Gentamicin and Amphotericin B 0.5 mL.
OcuLife™ Medium Complete Kit includes OcuLife™ Basal Medium and OcuLife™ LifeFactors® Kit. OcuLife™ Basal Medium requires the associated LifeFactors®.