Maxi Horizontal Gel System, Galileo Bioscience
Supplier: GeneMate
Certificates
About this item
GeneMate Large gels systems include "Express Cast" technology
Simply insert the gasketed end gates into the ends of the UV transparent gel tray, pour gel, remove combs and gasketed end gates, and run the gel. The 3-point leveling base with integrated bubble ensures gel system is level.
Ordering information: Includes Express Cast UV transparent gel tray with gasketed end gates, 3 x 1.5mm combs (1 x 16, 1 x 24, and 1 x 36-well).
Frequently Bought Together