Specifications
- Description:Adair Breast Tenaculum
- Length:6½"
- Material:Stainless steel
- Tip configuration:1x1 Prongs
- Curvature:Straight
- Cat. No.:10195-730
- Supplier no.:36-1865
Specifications
About this item
Sklar® Adair Breast Tenaculum Forceps are a ratcheted, finger ring instrument used to penetrate and grasp fibrocystic tissue or masses
They are perforating grasping forceps with sharp, non-crushing prong tips. This product has straight shanks with inward curving, single prong tips.