Small GTPase Agarose Beads, Cell Biolabs
Supplier: Cell Biolabs
Certificates
About this item
Cell Biolabs' Small GTPase Assay Beads are agarose beads containing a binding domain that selectively precipitates the active form of the small GTPase target of interest
The beads contain a light red color to allow easy visualization during the pulldown process. These are the same agarose beads found in Cell Biolabs' Small GTPase Activation Assays.
Specifications
- Storage Temperature:–20°C