Specifications
- Description:RhoA/Rac1/CDC42 Activation Assay Combo Kit
- Size:3 x 10 Assays
- Storage Temperature:–20°C
- Cat. No.:102965-128
- Method:Immunoblot
- Supplier no.:STA-405
Specifications
About this item
Cell Biolabs' RhoA/Rac1/Cdc42 Activation Assay Combo Kit uses visible agarose beads to selectively precipitate the active form of Rho, Rac and Cdc42
The precipitated small GTPase is then detected by Western blot using a RhoA-, Rac1- or Cdc42-specific antibody included in the kit. The assays use a safe, non-radioactive format. Each kit is sufficient to perform 10 assays for each protein.