ContactUsLinkComponent
Econo™ Tissue Nipper, Floor Grade, Sklar®
Econo™ Tissue Nipper, Floor Grade, Sklar®
Catalog # 89023-230
Supplier:  Sklar
undefined
Econo™ Tissue Nipper, Floor Grade, Sklar®
Catalog # 89023-230
Supplier:  Sklar
Supplier Number:  21-489
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Econo Nail Nipper
  • Length:
    4½"
  • Cat. No.:
    89023-230
  • Jaw type:
    Convex
  • Jaw length:
    11 mm
  • Supplier no.:
    21-489
Specifications
About this item
This is a double spring, plier handle instrument used for cutting soft tissue and cuticles from the nailbed.
  • Made of stainless steel
Econo™ instruments are Sklar's floor-grade, disposable instruments.