The QuickTiter™ Lentivirus Titer Kit (Lentivirus-Associated HIV p24) provides a unique and more accurate way to measure physical lentiviral titer
This kit substantially minimizes the problem of overestimating lentiviral titer with traditional p24 ELISA kits. The traditional p24 ELISA detects both virus-associated p24 and free p24 generated by 293 cells during transient transfection. Free p24 can account for a substantial portion of total p24 in the supernatant. Therefore, the ELISA typically overestimates the quantity of lentivirus present
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Conjugate ELISA:HRP
- Detection Method:Colorimetric
- Storage Temperature:4°C and –20°C
- Sensitivity:1 ng/mL