Order Entry
United States
ContactUsLinkComponent
ViraBind™ Adenovirus Miniprep Kit, Cell Biolabs
ViraBind™ Adenovirus Miniprep Kit, Cell Biolabs
Catalog # 102965-334
Supplier:  Cell Biolabs
CAS Number:  
undefined
ViraBind™ Adenovirus Miniprep Kit, Cell Biolabs
Catalog # 102965-334
Supplier:  Cell Biolabs
Supplier Number:  VPK-099
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Application:
  • Method / Format:
  • Refine target molecule:
  • Sample type:
  • Target molecule:
  • Description:
    Adenovirus Miniprep Kit
  • Size:
    10 Preps
  • Environmentally Preferable:
  • Storage Conditions:
    RT
  • Cat. No.:
    102965-334
  • Supplier no.:
    VPK-099

Specifications

About this item

ViraBind™ Adenovirus Purification Kits provide a much more efficient system for fast adenoviral purification with high yields, compared to the tedious, time-consuming CsCl procedure

The ViraBind™ Adenovirus Miniprep Kit uses a special spin column to purify supernatant from a single T75 flask or 10cm dish per prep in about 30 minutes. This kit achieves the quality of CsCl procedures in <2 hours, without ultracentrifugation, and allows >90% recovery rate.