OxiSelect™ Ferric Reducing Antioxidant Power (FRAP) Assay Kit, Cell Biolabs
Catalog # 102965-322
Supplier: Cell Biolabs
CAS Number:
Specifications
- Description:FRAP Assay Kit
- Size:200 Assays
- Storage Temperature:4°C
- Sensitivity:2 µM
- Cat. No.:102965-322
- Method:Colorimetric
- Supplier no.:STA-859
Specifications
About this item
OxiSelect™ Ferric Reducing Antioxidant Power (FRAP) Assay Kit is a quantitative assay for measuring the antioxidant potential within various samples
Following the reduction of ferric iron (Fe3+) to ferrous iron (Fe2+) by antioxidants present in the sample, the kit colorimetric probe develops a blue color that is read colorimetrically at 540-600nm. The antioxidant potential of samples is determined based on an iron standard curve and results are calculated at Fe2+ equivalents (µM) or FRAP value. This kit provides fast results after a quick 10 minute protocol, and it is suitable for use with serum, plasma, lysates, biological fluids, and purified food or drug extracts.