To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Assay Type:Competitive
- Conjugate ELISA:HRP
- Format:Uncoated
- Target protein:Nε-(carboxyethyl) lysine
- Description:CEL Competitive ELISA Kit
- Size:96 Assays
- Detection Method:Colorimetric
- Storage Temperature:4 °C and –20 °C
- Sensitivity:100 ng/mL
- Cat. No.:102965-282
- Supplier no.:STA-813
Specifications
About this item
OxiSelect™ Nε-(carboxyethyl) lysine (CEL) ELISA Kit provides rapid detection and quantitation of CEL protein adducts
First, a CEL conjugate is coated on the ELISA plate. The unknown CEL protein samples or CEL-BSA standards are then added to the CEL conjugate preabsorbed plate. After a brief incubation, the anti-CEL monoclonal antibody is added, followed by an HRP conjugated secondary antibody. The content of CEL protein adducts in unknown samples is determined by comparison with the predetermined CEL-BSA standard curve. Despite the structure similarity between CEL and CML, the anti-CEL specific antibody in the OxiSelect™ CEL ELISA Kit will not cross react with CML protein adducts.