To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Description:Glutathione Reductase Assay Kit
- Size:100 Assays
- Storage Temperature:4°C and –80°C
- Sensitivity:0.6 mU/mL
- Cat. No.:102965-280
- Method:Colorimetric
- Supplier no.:STA-812
OxiSelect™ Glutathione Reductase Assay Kit is a quantitative assay for measuring glutathione reductase activity within plasma, erythrocytes, tissues, and cell lysates
Glutathione reductase activity is defined as 1 unit of enzyme reducing 1 µmole oxidized glutathione (GSSG) per minute at pH 7.6 and 25ºC. The kit employs a simple enzymatic recycling reaction for glutathione quantification where the reduction of a chromagen is correlated to glutathione reductase enzymatic activity. The kit can detect glutathione reductase activity as low as 0.6 mU/mL.