To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:pWZL-Neo Retroviral Vector
- Size:10 μg
- Environmentally Preferable:
- Format:Purified Plasmid
- Storage Temperature:–20°C
- Cat. No.:102964-496
- Supplier no.:RTV-001-NEO
Specifications
About this item
Cell Biolabs' pWZL vector is suitable for making retrovirus for general gene expression in various cells
Clone your gene of interest into this vector and then transfect the construct into a retroviral packaging cell line, 293T cells, or 293RTV cells. This vector contains a neomycin resistance gene.