Order Entry
United States
ContactUsLinkComponent
Lactose Assay Kit, Cell Biolabs
Lactose Assay Kit, Cell Biolabs
Catalog # 102966-204
Supplier:  Cell Biolabs
CAS Number:  
undefined
Lactose Assay Kit, Cell Biolabs
Catalog # 102966-204
Supplier:  Cell Biolabs
Supplier Number:  MET-5001
CAS Number:  

Specifications

  • Description:
    Lactose Assay Kit
  • Size:
    100 Assays
  • Storage Temperature:
    Room Temperature and –20°C
  • Sensitivity:
    10µM
  • Cat. No.:
    102966-204
  • Method:
    Fluorometric
  • Supplier no.:
    MET-5001

Specifications

About this item

Cell Biolabs' Lactose Assay Kit measures total lactose in milk based food products or biological samples (such as blood or urine from lactating mammals)

Lactose is first cleaved into glucose and galactose by lactase. Glucose is then oxidized by glucose oxidase, yielding hydrogen peroxide and D-gluconic acid. The hydrogen peroxide released from this reaction is detected by a fluorometric probe in a 1:1 ratio. Lactose levels in unknown samples are determined based on a lactose standard curve. The Lactose Assay Kit can detect lactose concentrations as low as 10µM.