PYREX® Claisen Three-Way Connecting Adapter, with 19/22 [ST] Joints, Corning
Catalog # 32645-367
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Description:PYREX® Claisen Three-Way Connecting Adapter, with [ST] Joints
- Environmentally Preferable:Low Manufacturing Impact
- Environmental Certification:I-RECs, ISO 14001 - Monterrey
- Cat. No.:32645-367
- [ST] joint size:19/22
- Supplier no.:9050-19
Specifications
About this item
These PYREX® Claisen three-way adapters have standard taper outer joints at the upper end and on the parallel sidearm, while an inner standard taper joint is positioned at the lower end.
- Top and sidearm [ST] outer joints
- Bottom [ST] inner joint
Adapters feature 19/22 standard saper outer joints at the upper end and on the parallel sidearm. An inner 19/22 standard taper joint is positioned at the lower end.