ContactUsLinkComponent
PYREX® Capillary Tip Bleed Tube, Corning
PYREX® Capillary Tip Bleed Tube, Corning
Catalog # 26658-037
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
undefined
PYREX® Capillary Tip Bleed Tube, Corning
Catalog # 26658-037
Supplier:  Corning
Supplier Number:  7815-19
Product available on GSA Advantage®
   Low Manufacturing Impact
Specifications
  • Description:
    PYREX® Capillary Tip Bleed Tube, 280 mm
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental Certification:
    I-RECs, ISO 14001 - Monterrey
  • Cat. No.:
    26658-037
  • Supplier no.:
    7815-19
Specifications
About this item
For bleeding gases, steam, or liquids below the liquid surface of sidearm distillation flasks.