About this item
Dulbecco’s Modifed Eagle's medium (DMEM) is a modification of Basal Medium Eagle (BME) which contains approximately four times as much of the vitamins and amino acids and two to four times as much glucose as the original formulation.
- Suitable for most types of cells
- Available in a variety of formulations
- Available with or without sodium pyruvate
- Available with or without L- glutamine
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Related Information
Certificates
Search by Lot Number02-0100 spec DMEM 4.5gL glucose, wout sod pyr w l-glut 02-0101 spec DMEM 4.5gL glucose, w sod pyr w l-glut 02-0102 spec DMEM 4.5gL glucose, wout sod pyr wout l-glut 02-0111 spec DMEM 1.0gL glucose, w sod pyr w l-glut 02-0148 spec DMEM 4.5gL glucose, w sod pyr wout l-glut
Datasheet
Focus Industrial VWR bioMarke Magazine Volume 5 Issue 1 VWR Essential Products for Tissue Culture VWR_Life_Science_Dulbeccos_Modified_Eagle_Medium_DMEM_Formulation_Sheet VWRbioMarke Advancing The Cell
Product Literature
spec_VWRL0100-0500 spec_VWRL0101-0500 spec_VWRL0102-0500 spec_VWRL0111-0500 spec_VWRL0111-0500