OxiSelect™ Cellular UV-Induced DNA Damage ELISA Kit mesaures the formation of cyclobutane pyrimidine dimers (CPD) in intact cells
Cells are first seeded in a 96-well tissue culture plate. Wells are then UV irradiated to induce DNA damage. After fixation and denaturation, cells containing the DNA lesions are probed with an anti-CPD antibody, followed by an HRP conjugated secondary antibody.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Detection Method:Colorimetric
- Storage Temperature:4°C
- Conjugate ELISA:HRP