OxiSelect™ UV-Induced DNA Damage ELISA Kit measures 6-4PP formation in DNA isolated from cells or tissue
Standards or unknown DNA samples are first heat denatured before adsorbed onto a 96-well DNA binding plate. The sample or standard is then probed with an anti-6-4PP antibody, followed by an HRP conjugated secondary antibody.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Conjugate ELISA:HRP
- Detection Method:Colorimetric
- Storage Temperature:4°C and –20°C