OxiSelect™ Oxidative DNA Damage ELISA Kit (8-OHdG Quantitation), Cell Biolabs
Supplier: Cell Biolabs
Certificates
About this item
OxiSelect™ Oxidative DNA Damage ELISA Kit (8-hydroxydeoxyguanosine assay) provides a powerful method for rapid, sensitive quantitation of 8-OHdG in DNA samples
This kit is suitable for use with urine, serum, cells or tissues. Detect as little as 100 pg/mL of 8-OHdG. An 8-OHdG standard included for absolute quantitation.
Among numerous types of oxidative DNA damage, 8-hydroxydeoxyguanosine (8-OHdG) is a ubiquitous marker of oxidative stress. 8-OHdG, one of the byproducts of oxidative DNA damage, is physiologically formed and enhanced by chemical carcinogens.
Specifications
- Conjugate ELISA:HRP
- Detection Method:Colorimetric
- Storage Temperature:4°C, –20°C and –80°C
- Sensitivity:100 pg/mL
Specifications
Frequently Bought Together