Anti-CEBPA Rabbit Polyclonal Antibody
Catalog # 10813-794
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:CCAAT/enhancer binding protein (C/EBP), alpha
- Antigen Symbol:CEBPA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:0.1 mg
- Form:Liquid
- Gene ID:P49715
- Antigen Synonyms:CEBPA
- Modification:Thr 230
- Storage Buffer:Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
- Molecular Weight:42 kDa
- Storage Temperature:Store antibody at –20 °C for up to one year.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:C/EBP-alpha (phospho Thr230) antibody was raised against a peptide sequence around phosphorylation site of threonine 230 (P-P-T (p)-P-V) derived from Human C/EBP-alpha.
- Purification:Antibodies were purified by affinity-chromatography using epitope-specific peptide.
- Cat. No.:10813-794
- Supplier no.:79-898
Specifications
About this item
The protein encoded by this intronless gene is a bZIP transcription factor which can bind as a homodimer to certain promoters and enhancers. It can also form heterodimers with the related proteins CEBP-beta and CEBP-gamma. The encoded protein has been shown to bind to the promoter and modulate the expression of the gene encoding leptin, a protein that plays an important role in body weight homeostasis. Also, the encoded protein can interact with CDK2 and CDK4, thereby inhibiting these kinases and causing growth arrest in cultured cells.
Western Blot: 1:500~1:1000
Type: Primary
Antigen: CEBPA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat