- Antibody Type:Primary
- Antigen Name:discoidin domain receptor tyrosine kinase 1
- Antigen Symbol:DDR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:0.1 mg
- Form:Liquid
- Gene ID:Q08345
- Antigen Synonyms:DDR1
- Modification:Tyr 513
- Storage Buffer:Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
- Molecular Weight:110 kDa
- Storage Temperature:Store antibody at –20 °C for up to one year.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:DDR1 (phospho Tyr513) antibody was raised against a peptide sequence around phosphorylation site of tyrosine 513 (K-K-Y (p)-V-R) derived from Human DDR1 .
- Purification:Antibodies were purified by affinity-chromatography using epitope-specific peptide.
- Cat. No.:10813-760
- Supplier no.:79-881
Receptor tyrosine kinases (RTKs) play a key role in the communication of cells with their microenvironment. These molecules are involved in the regulation of cell growth, differentiation and metabolism. The protein encoded by this gene is a RTK that is widely expressed in normal and transformed epithelial cells and is activated by various types of collagen. This protein belongs to a subfamily of tyrosine kinase receptors with a homology region to the Dictyostelium discoideum protein discoidin I in their extracellular domain. Its autophosphorylation is achieved by all collagens so far tested (type I to type VI).
Western Blot: 1:500~1:1000, Immunohistochemistry: 1:50~1:100
Type: Primary
Antigen: DDR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat