OxiSelect™ Protein Carbonyl Fluorometric Assay, Cell Biolabs
Catalog # 102964-868
Supplier: Avantor
CAS Number:
Specifications
- Description:Protein Carbonyl Fluorometric Assay
- Size:100 Assays
- Storage Temperature:4°C and –20°C
- Cat. No.:102964-868
- Method:Fluorometric
- Supplier no.:STA-307
Specifications
About this item
OxiSelect™ Protein Carbonyl Fluorometric Assay Kit provides a rapid, efficient method for the detection of protein carbonyl residues
The carbonyl derivatives of proline, lysine, arginine and threonine residues are the most common products of protein oxidation in biological samples. Such derivatives are chemically stable and serve as markers for oxidative stress in most types of reactive oxygen species. The fluorescence plate-based format provides a convenient system for direct measurement of protein carbonyl content. The kit is suitable for plasma, serum, urine, cell lysates or purified proteins.