OxiSelect™ Nitrotyrosine ELISA Kit, Cell Biolabs
OxiSelect™ Nitrotyrosine ELISA Kit provides a simple and sensitive method to measure the formation of 3-nitrotyrosine in proteins
Nitrotyrosine serves as a downstream marker for peroxynitrite and nitric oxide in a variety of disease states and in conditions of cellular damage and oxidative stress. Detect nitrotyrosine concentrations as low as 10 nM. The kit is suitable for use with cell lysates, serum, plasma and purified proteins.
- Conjugate ELISA:HRP
- Detection Method:Colorimetric
- Storage Temperature:4°C and –20°C
- Sensitivity:10 nM
Frequently Bought Together