About this item
AAV Helper Free Packaging Systems contain the following constructs needed to package your recombinant adeno-associated virus along with your own expression vector: pHelper plasmid, a serotype-specific Rep-Cap plasmid, and an optional GFP expression vector for making a control virus
These systems are useful if you already have a cloning vector or expression construct containing your gene of interest. AAV Helper Free Packaging Systems are available for native serotypes 1 through 6
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Format:Purified Plasmid
- Storage Temperature:–20°C