Order Entry
United States
ContactUsLinkComponent
 
HDL-Cholesterol Assay Kit, Cell Biolabs
Catalog # 102965-076
Supplier:  Cell Biolabs
CAS Number:  
undefined
HDL-Cholesterol Assay Kit, Cell Biolabs
Catalog # 102965-076
Supplier:  Cell Biolabs
Supplier Number:  STA-394
CAS Number:  

Specifications

  • Description:
    HDL-Cholesterol Assay Kit
  • Size:
    96 Assays
  • Storage Temperature:
    4°C and –20°C
  • Cat. No.:
    102965-076
  • Method:
    Fluorometric
  • Supplier no.:
    STA-394

Specifications

About this item

Cell Biolabs’ HDL-Cholesterol Assay Kit is a simple fluorometric assay that can measure the amounts of HDL cholesterol present in plasma or serum samples within a 96-well microtiter plate format.  Each kit provides sufficient reagents to perform up to 96 assays, including blanks, cholesterol standards and unknown samples.  Sample cholesterol concentrations are determined by comparison with a known cholesterol standard.