Human LOX-1 ELISA Kit, Cell Biolabs
Catalog # 102965-062
Supplier: Cell Biolabs
CAS Number:
Specifications
- Primary antibody reactivity:Human
- Description:Human LOX-1 ELISA Kit
- Size:96 Assays
- Storage Conditions:4°C and –20°C
- Sensitivity:40 pg/mL
- Cat. No.:102965-062
- Supplier no.:STA-387
Specifications
About this item
Cell Biolabs’ Human LOX-1 ELISA Kit is an enzyme immunoassay developed for the detection and quantitation of human LOX-1 in plasma, serum or cell culture supernatant samples
The Human LOX-1 ELISA Kit has a detection sensitivity limit of 40 pg/mL human LOX-1. Each kit provides sufficient reagents to perform up to 96 assays including standard curve and unknown samples.
The lectin-like oxidized LDL receptor-1 (LOX-1) is a 50 kDa type II transmembrane glycoprotein that structurally belongs to the C-type lectin family. LOX-1 is expressed in vascular endothelium, vascular smooth muscle cells, differentiated macrophages and platelets.