To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Description:Cell Viability and Cytotoxicity Assay
- Size:96 Assays
- Storage Temperature:4°C and –20°C
- Cat. No.:102963-842
- Method:Colorimetric/fluorometric
- Supplier no.:CBA-240
The CytoSelect™ Cell Viability and Cytotoxicity Assay Kit provides a simple format for monitoring cell viability via metabolic activity
Prior to the assay, cells may be treated with compounds or agents that affect cell viability. This kit is suitable for eukaryotic cells, but not yeast or bacteria.
Live cells are detected with either MTT (colorimetric detection) or Calcein AM (fluorometric detection). Dead cells are detected by EthD-1 reagent (fluorometric). All 3 detection reagents are included, along with Saponin (a cell death initiator).