Order Entry
United States
Orders LinkContactUsLinkComponent
Envelope Vectors, Cell Biolabs

Envelope Vectors, Cell Biolabs

Marketsource
Supplier:   Cell Biolabs
Envelope Vectors, Cell Biolabs
Supplier:  Cell Biolabs
Marketsource
Certificates
Envelope Vectors, Cell Biolabs

1 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Cell Biolabs' Envelope Vectors each contain a different envelope gene for pseudotyping recombinant lentivirus or retrovirus

Co-transfect one of these constructs along with other packaging and cloning vectors into 293T or 293RTV cells.

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 102965-756
  • 102965-760
  • 102965-762
  • 102965-764
  • Format:
  • Purified Plasmid
  • Bacterial Glycerol Stock
  • Bacterial Glycerol Stock
  • Bacterial Glycerol Stock
  • Storage Temperature:
  • –20°C
  • –80°C
  • –80°C
  • –80°C

Specifications

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 102965-756
    pCMV-VSV-G
    pCMV-VSV-G
    10 μg
    Log in for availability
    Log in for availability
    Each
    Each - $763.08
    $763.08
    Each
       $763.08
    Minimum order quantity is 1

    Supplier Number: RV-110
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    102965-756
    Format
    Purified Plasmid
    Storage Temperature
    –20°C
    Marketsource
  • 102965-760
    pCMV-Eco
    pCMV-Eco
    100 µL
    Discontinued
    Supplier Number: -
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    102965-760
    Format
    Bacterial Glycerol Stock
    Storage Temperature
    –80°C
    Marketsource
  • 102965-762
    pCMV-Ampho
    pCMV-Ampho
    100 µL
    Discontinued
    Supplier Number: -
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    102965-762
    Format
    Bacterial Glycerol Stock
    Storage Temperature
    –80°C
    Marketsource
  • 102965-764
    pCMV-10A1
    pCMV-10A1
    100 µL
    Discontinued
    Supplier Number: -
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    102965-764
    Format
    Bacterial Glycerol Stock
    Storage Temperature
    –80°C
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.