Order Entry
United States
ContactUsLinkComponent
Anti-IgG Goat Polyclonal Antibody (IRDye® 800CW)
Anti-IgG Goat Polyclonal Antibody (IRDye® 800CW)
Catalog # 103011-494
Supplier:  Li-Cor
Anti-IgG Goat Polyclonal Antibody (IRDye® 800CW)
Catalog # 103011-494
Supplier:  Li-Cor
Supplier Number:  925-32211

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Secondary
  • Antigen Name:
    Immunoglobulin G
  • Antigen Symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    IRDye® 800CW
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Rabbit
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Cross Adsorption:
    Highly Cross-adsorbed
  • Form:
    Lyophilized
  • Storage Buffer:
    PBS, pH 7.4. Contains 10 mg/mL BSA (free of IgG and protease) as stabilizer and 0.1% sodium azide as preservative, after reconstitution
  • Storage Temperature:
    Protect from light. Store at 4 °C prior to reconstitution
  • Tested Applications:
    On-Cell Western Assay, Protein Array, Microscopy, 2D Gel Detection, Tissue Section Imaging, Small Animal Imaging, Virus Titration, FRET-based Assay = Yes
  • Cat. No.:
    103011-494
  • Supplier no.:
    925-32211

Specifications

About this item

Isolation of specific antibodies was accomplished by affinity chromatography using pooled rabbit IgG covalently linked to agarose beads. Based on ELISA and flow cytometry, this antibody reacts with the heavy and light chains of rabbit IgG, and with the light chains of rabbit IgM and IgA. This antibody was tested by Dot Blot and/or solid-phase adsorbed for minimal cross-reactivity with human, mouse, rat, sheep, and chicken serum proteins, but may cross-react with immunoglobulins from other species. The conjugate has been specifically tested and qualified for Western blot and In-Cell Western™ assay applications. Highly Recommended for: Western blot, In-Cell Western Assay, On-Cell Western Assay, protein array, immunohistochemistry, microsciopy, 2D gel detection, tissue section imaging, virus titration assay, FRET-based assays, and small animal imaging.

Western blot: 1:5000

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: IRDye 800CW
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit