Order Entry
United States
ContactUsLinkComponent
Anti-IgG Donkey Polyclonal Antibody (IRDye® 800CW)
Anti-IgG Donkey Polyclonal Antibody (IRDye® 800CW)
Catalog # 102970-994
Supplier:  Li-Cor
Anti-IgG Donkey Polyclonal Antibody (IRDye® 800CW)
Catalog # 102970-994
Supplier:  Li-Cor
Supplier Number:  925-32213

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Secondary
  • Antigen Name:
    Immunoglobulin G
  • Antigen Symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    IRDye® 800CW
  • Host:
    Donkey
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Rabbit
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Cross Adsorption:
    Highly Cross-adsorbed
  • Form:
    Lyophilized
  • Storage Buffer:
    PBS, pH 7.4. Contains 10 mg/mL BSA (free of IgG and protease) as stabilizer and 0.1% sodium azide as preservative, after reconstitution
  • Storage Temperature:
    Protect from light. Store at 4 °C prior to reconstitution
  • Tested Applications:
    On-Cell Western Assay, Protein Array, Microscopy, 2D Gel Detection, Tissue Section Imaging, Small Animal Imaging, Virus Titration, FRET-based Assay = Yes
  • Cat. No.:
    102970-994
  • Supplier no.:
    925-32213

Specifications

About this item

The antibody was isolated by affinity chromatography using antigens coupled to agarose beads. Based on ELISA, this antibody reacts with the heavy and light chains on rabbit IgG and with the light chains common to most rabbit immunoglobulins. This antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reactivity with bovine, chicken, goat, guinea pig, hamster, horse, human, mouse, rat, and sheep serum proteins, but the antibody may cross-react with immunoglobulins from other species. The conjugate has been specifically tested and qualified for Western blot and In-Cell Western™ assay applications. Highly Recommended for: Western blot, In-Cell Western Assay, On-Cell Western Assay, protein array, immunohistochemistry, microsciopy, 2D gel detection, tissue section imaging, virus titration assay, FRET-based assays, and small animal imaging.

Western blot: 1:5000

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: IRDye 800CW
Epitope:
Host: Donkey
Isotype:
Reactivity: Rabbit