Order Entry
United States
ContactUsLinkComponent
Anti-IgG Donkey Polyclonal Antibody (IRDye® 800CW)
Anti-IgG Donkey Polyclonal Antibody (IRDye® 800CW)
Catalog # 102673-358
Supplier:  Li-Cor
Anti-IgG Donkey Polyclonal Antibody (IRDye® 800CW)
Catalog # 102673-358
Supplier:  Li-Cor
Supplier Number:  926-32411

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Secondary
  • Antigen Name:
    Immunoglobulin G
  • Antigen Symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    IRDye® 800CW
  • Host:
    Donkey
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Guinea pig
  • Western Blot:
    Yes
  • Size:
    500 µg
  • Cross Adsorption:
    Highly Cross-adsorbed
  • Form:
    Lyophilized
  • Storage Buffer:
    PBS, pH 7.4. Contains 10 mg/mL BSA (free of IgG and protease) as stabilizer and 0.1% sodium azide as preservative, after reconstitution
  • Storage Temperature:
    Protect from light. Store at 4 °C prior to reconstitution
  • Tested Applications:
    On-Cell Western Assay, Protein Array, Microscopy, 2D Gel Detection, Tissue Section Imaging, Small Animal Imaging, Virus Titration, FRET-based Assay = Yes
  • Cat. No.:
    102673-358
  • Supplier no.:
    926-32411

Specifications

About this item

Isolation of specific antibodies was accomplished by affinity chromatography using antigens immobilized on agarose beads. Based on immunoelectrophoresis and/or ELISA, this antibody reacts with whole molecule guinea pig IgG as well as the light chains common to other guinea pig immunoglobulins. No reactivity was detected against non-immunoglobulin serum proteins. This antibody was tested by ELISA and/or solid phase adsorbed for minimal cross-reactivity with bovine, chicken, goat, Syrian hamster, horse, human, mouse, rabbit, rat, and sheep serum proteins, but may cross-react with immunoglobulins from other species. The conjugate has been specifically tested and qualified for Western blot and In-Cell Western™ assay applications. Highly Recommended for: Western blot, In-Cell Western Assay, On-Cell Western Assay, protein array, immunohistochemistry, microsciopy, 2D gel detection, tissue section imaging, virus titration assay, FRET-based assays, and small animal imaging.

Western blot: 1:5000

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: IRDye 800CW
Epitope:
Host: Donkey
Isotype:
Reactivity: Guinea Pig