Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin G
- Antigen Symbol:IgG
- Clonality:Polyclonal
- Conjugation:IRDye® 800CW
- Host:Donkey
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Chicken
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:Highly Cross-adsorbed
- Form:Lyophilized
- Storage Buffer:PBS, pH 7.4. Contains 10 mg/mL BSA (free of IgG and protease) as stabilizer and 0.1% sodium azide as preservative, after reconstitution
- Storage Temperature:Protect from light. Store at 4 °C prior to reconstitution
- Tested Applications:On-Cell Western Assay, Protein Array, Microscopy, 2D Gel Detection, Tissue Section Imaging, Small Animal Imaging, Virus Titration, FRET-based Assay = Yes
- Cat. No.:102970-998
- Supplier no.:925-32218
Specifications
About this item
The antibody was isolated from antisera by affinity chromatography using antigens coupled to agarose beads. Based on immunoelectrophoresis and/or ELISA, the antibody reacts with whole molecule chicken IgY. It also reacts with the light chains of other chicken immunoglobulins. No reactivity was detected against non-immunoglobulin serum proteins. This antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reactivity with bovine, goat, guinea pig, Syrian hamster, horse, mouse, human, rabbit, rat, and sheep serum proteins, but the antibody may cross-react with immunoglobulins from other species. The conjugate was tested and qualified for Western blot applications. Highly Recommended for: Western blot, In-Cell Western Assay, On-Cell Western Assay, protein array, immunohistochemistry, microsciopy, 2D gel detection, tissue section imaging, virus titration assay, FRET-based assays, and small animal imaging.
Western blot: 1:5000
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: IRDye 800CW
Epitope:
Host: Donkey
Isotype:
Reactivity: Chicken