Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin G
- Antigen Symbol:IgG
- Clonality:Polyclonal
- Conjugation:IRDye® 680RD
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:Highly Cross-adsorbed
- Form:Lyophilized
- Storage Buffer:PBS, pH 7.4. Contains 10 mg/mL BSA (free of IgG and protease) as stabilizer and 0.1% sodium azide as preservative, after reconstitution
- Storage Temperature:Protect from light. Store at 4 °C prior to reconstitution
- Tested Applications:On-Cell Western Assay, Protein Array, Microscopy, 2D Gel Detection, Tissue Section Imaging, Small Animal Imaging = Yes
- Cat. No.:102971-034
- Supplier no.:925-68078
Specifications
About this item
Isolation of specific antibodies was accomplished by affinity chromatography using human IgG covalently linked to agarose beads. Based on ELISA, the antibody reacts with the heavy and light chains of human IgG. This antibody was tested by Dot Blot and/or solid-phase adsorbed for minimal cross-reactivity with bovine, horse, and mouse serum proteins, but may cross-react with immunoglobulins from other species. The conjugate has been specifically tested and qualified for Western blot and In-Cell Western™ assay applications. Highly Recommended for: Western blot, In-Cell Western Assay, On-Cell Western Assay, protein array, immunohistochemistry, microsciopy, 2D gel detection, tissue section imaging, and small animal imaging.
Western blot: 1:15000; In-Cell Western Assay: 1:800
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: IRDye 680RD
Epitope:
Host: Goat
Isotype:
Reactivity: Human