Order Entry
United States
ContactUsLinkComponent
Anti-IgM Goat Polyclonal Antibody (IRDye® 680LT)
Anti-IgM Goat Polyclonal Antibody (IRDye® 680LT)
Catalog # 102971-036
Supplier:  Li-Cor
Anti-IgM Goat Polyclonal Antibody (IRDye® 680LT)
Catalog # 102971-036
Supplier:  Li-Cor
Supplier Number:  925-68080

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Secondary
  • Antigen Name:
    Immunoglobulin M
  • Antigen Symbol:
    IgM
  • Clonality:
    Polyclonal
  • Conjugation:
    IRDye® 680LT
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Cross Adsorption:
    Highly Cross-adsorbed
  • Form:
    Lyophilized
  • Storage Buffer:
    PBS, pH 7.4. Contains 10 mg/mL BSA (free of IgG and protease) as stabilizer and 0.1% sodium azide as preservative, after reconstitution
  • Storage Temperature:
    Protect from light. Store at 4 °C prior to reconstitution
  • Tested Applications:
    Protein Array, Microscopy, 2D Gel Detection, Tissue Section Imaging,
  • Cat. No.:
    102971-036
  • Supplier no.:
    925-68080

Specifications

About this item

Isolation of specific antibodies was accomplished by affinity chromatography using mouse IgM covalently linked to agarose beads. Based on ELISA and/or flow cytometry, this antibody reacts with the heavy chain of mouse IgM. This antibody was tested by Dot Blot and/or solid phase adsorbed for minimal cross-reactivity with mouse IgG1, IgG2a, IgG3, and IgA, pooled human sera and purified human paraproteins. The conjugate has been specifically tested and qualified for Western blot. The conjugate has been specifically tested and qualified for Western blot applications. Recommended for: Western blot, proein array, immunohistochemistry, microscopy, 2D gel detection, and tissue section imaging. NOT RECOMMENDED for: small animal imaging, In-Cell Wesern assays, or On-Cell Western assays.

Western blot: 1:20000

Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: IRDye 680LT
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse