Collagen-based Cell Contraction Assay, Cell Biolabs
Catalog # 102963-828
Supplier: Cell Biolabs
CAS Number:
Specifications
- Description:Cell Contraction Assay
- Size:24 Assays
- Storage Temperature:4°C
- Cat. No.:102963-828
- Method:Light Microscopy
- Supplier no.:CBA-201
Specifications
About this item
Cell Biolabs’ Collagen-based Contraction Assay Kit provides a simple system to assess cell contractivity in vitro and screen cell contraction mediators
Each kit provides sufficient quantities to perform up to 24 assays in a 24-well plate. The kit can also be used for culturing cells in a 3D collagen matrix.