CytoSelect™ Clonogenic Tumor Cell Isolation Kit, Cell Biolabs
Supplier: Cell Biolabs
Certificates
About this item
CytoSelect™ Clonogenic Tumor Cell Isolation Kit uses a proprietary semisolid agar medium to facilitate formation of colonies by cells from solid tumors
The kit allows separation of neoplastic colony-forming cells from non-clonogenic cells in heterogeneous tumor samples. Colonies are grown in either a 6-well plate or a 35mm culture dish. These colonies are isolated away from single (i.e. normal) cells by size filtration. The viable cells from these colonies can be easily recovered for further analysis.
Specifications
- Storage Temperature:4°C