Radius™ Cell Migration Assay Kits, Cell Biolabs
About this item
Radius™ Cell Migration Assays provide a convenient and user-friendly method to measure two-dimensional cell migration
Unlike traditional Boyden Chamber assays, each Radius plate can accept cells of any type and size without the worry of choosing a pore sized matched to your specific cell. Kits are available in 24-well, 96-well, and 384-well formats. For cells that need an ECM protein for stronger cell adhesion, we offer pre-coated 24-well plates with Collagen I, Fibronectin or Laminin. We also provide an ECM Array kit which contains a 24-well plate with one row of each aforementioned protein plus one row uncoated.
When cells are seeded in the well of the Radius plate, they will adhere everywhere except in the center of the well where we have placed a biocompatible hydrogel spot. Once cells form a monolayer, the assay is initiated by gently dissolving the gel with a removal solution, leaving a gap across which cell migration can occur.
4 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Radius™ Cell Migration Assays provide a convenient and user-friendly method to measure two-dimensional cell migration
Unlike traditional Boyden Chamber assays, each Radius plate can accept cells of any type and size without the worry of choosing a pore sized matched to your specific cell. Kits are available in 24-well, 96-well, and 384-well formats. For cells that need an ECM protein for stronger cell adhesion, we offer pre-coated 24-well plates with Collagen I, Fibronectin or Laminin. We also provide an ECM Array kit which contains a 24-well plate with one row of each aforementioned protein plus one row uncoated.
When cells are seeded in the well of the Radius plate, they will adhere everywhere except in the center of the well where we have placed a biocompatible hydrogel spot. Once cells form a monolayer, the assay is initiated by gently dissolving the gel with a removal solution, leaving a gap across which cell migration can occur.
Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 102963-772
- 102963-774
- 102963-776
- 102963-778
- 102963-780
- 102963-782
- 102963-784
- 102963-786
- 102963-788
- 102963-790
- Storage Temperature:
- Room Temperature, 4°C and –20°C
- Room Temperature, 4°C and –20°C
- Room Temperature, 4°C and –20°C
- Room Temperature, 4°C and –20°C
- Room Temperature, 4°C and –20°C
- Room Temperature, 4°C and –20°C
- Room Temperature, 4°C and –20°C
- Room Temperature, 4°C and –20°C
- Room Temperature, 4°C and –20°C
- Room Temperature, 4°C and –20°C
Specifications
Product Family Options
Product Information
- SizeMethodAvailabilityPrice
- 102963-77224-Well AssaySupplier Number: CBA-125Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:Marketsource
More
Specifications
Cat. No.102963-772Storage TemperatureRoom Temperature, 4°C and –20°CMarketsource - 102963-77424-Well AssaySupplier Number: CBA-125-5Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:Marketsource
More
Specifications
Cat. No.102963-774Storage TemperatureRoom Temperature, 4°C and –20°CMarketsource - 102963-77624-Well Assay, Collagen I Coated24-Well Assay, Collagen I Coated24 AssaysMicroscopyDiscontinued-Supplier Number: -Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:Marketsource
More
Specifications
Cat. No.102963-776Storage TemperatureRoom Temperature, 4°C and –20°CMarketsource - 102963-77824-Well Assay, ECM Array24-Well Assay, ECM Array24 AssaysMicroscopyDiscontinued-Supplier Number: CBA-125-ECMItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:Marketsource
More
Specifications
Cat. No.102963-778Storage TemperatureRoom Temperature, 4°C and –20°CMarketsource - 102963-78024-Well Assay, Fibronectin Coated24-Well Assay, Fibronectin Coated24 AssaysMicroscopyDiscontinued-Supplier Number: CBA-125-FNItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:Marketsource
More
Specifications
Cat. No.102963-780Storage TemperatureRoom Temperature, 4°C and –20°CMarketsource - 102963-78224-Well Assay, Laminin Coated24-Well Assay, Laminin Coated24 AssaysMicroscopyDiscontinued-Supplier Number: -Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:Marketsource
More
Specifications
Cat. No.102963-782Storage TemperatureRoom Temperature, 4°C and –20°CMarketsource - 102963-78496-Well AssaySupplier Number: CBA-126Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:Marketsource
More
Specifications
Cat. No.102963-784Storage TemperatureRoom Temperature, 4°C and –20°CMarketsource - 102963-78696-Well AssaySupplier Number: CBA-126-5Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:Marketsource
More
Specifications
Cat. No.102963-786Storage TemperatureRoom Temperature, 4°C and –20°CMarketsource - 102963-788384-Well Assay384-Well Assay384 AssaysMicroscopyDiscontinued-Supplier Number: -Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:Marketsource
More
Specifications
Cat. No.102963-788Storage TemperatureRoom Temperature, 4°C and –20°CMarketsource - 102963-790384-Well Assay384-Well Assay5 x 384 wellsMicroscopyDiscontinued-Supplier Number: -Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:Marketsource
More
Specifications
Cat. No.102963-790Storage TemperatureRoom Temperature, 4°C and –20°CMarketsource